accessibility menu, dialog, popup

UserName

CAS RN 147-14-8

IUPAC Name: 9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Synonyms: pigment blue 15:1
Molecular Weight (g/mol): 576.08
Molecular Formula: C32H16CuN8
InChi Key: XCJYREBRNVKWGJ-UHFFFAOYSA-N
SMILES: [Cu]1N2C3=NC4=NC(=NC5=C6C=CC=CC6=C(N=C6N=C(N=C2C2=CC=CC=C32)C2=CC=CC=C62)N15)C1=CC=CC=C41
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (5)
  • (3)
Color 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (5)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)
  • (2)