missing translation for 'onlineSavingsMsg'
Learn More

Copper phthalocyanine, dye content ca 95%

Catalog No. AC269152500
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269152500 250 g Glass bottle
AC269150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269152500 Supplier Thermo Scientific Chemicals Supplier No. 269152500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 147-14-8 | C32H16CuN8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160, Pigment blue 15

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Merck Index 15, 2505
Color Blue
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons
Quantity 250 g
Physical Form Fine Crystalline Powder
Formula Weight 576.08
Percent Purity ≥94% (ex Cu)
Chemical Name or Material Copper phthalocyanine, dye content ca 95%
Show More Show Less
EINECSNumber 205-685-1
RTECSNumber GL8510000
TSCA TSCA

RUO – Research Use Only