missing translation for 'onlineSavingsMsg'
Learn More

Pigment Blue 15 (purified by sublimation) 98.0+%, TCI America™

Catalog No. P16281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16281G Supplier TCI America Supplier No. P16281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym Phthalocyanine Blue, Copper Phthalocyanine, CuPc

Specifications

Melting Point 480°C
Color Blue/Purple
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 576.08
Percent Purity ≥98.0% (T)
Chemical Name or Material Pigment Blue 15 (purified by sublimation)
EINECSNumber (5)-3299&(5)-5216
RTECSNumber GL8510000
TSCA Yes