accessibility menu, dialog, popup

UserName

CAS RN 1465-25-4

IUPAC Name: dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Synonyms: dihydrogen N-(1-naphthyl)ethylenediamine dichloride
Molecular Weight (g/mol): 259.17
Molecular Formula: C12H16Cl2N2
InChi Key: MZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (8)
  • (5)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (13)
  • (14)
  • (1)
Grade 
  • (9)
  • (4)
  • (4)
  • (6)
Physical Form 
  • (3)
  • (1)
  • (8)
  • (9)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (8)
  • (1)
  • (2)
  • (1)
  • (6)
  • (8)
  • (3)
  • (9)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (8)
  • (5)
  • (1)
  • (1)
  • (2)
  • (13)
  • (14)
  • (1)
  • (9)
  • (4)
  • (4)
  • (6)
  • (3)
  • (1)
  • (8)
  • (9)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)