missing translation for 'onlineSavingsMsg'
Learn More

Naphthylethylenediamine Dihydrochloride, 0.1% (w/v), Ricca Chemical

Catalog No. 52304
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
52304 120 mL Amber Glass Bottle
523016 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52304 Supplier Ricca Chemical Company Supplier No. 52304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA for Nitrite Nitrogen Testing

Chemical Identifiers

Molecular Formula C12H16Cl2N2
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N

Specifications

Color Brown to Green-Yellow
CAS 1465-25-4, 7732-18-5
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.174
PubChem CID 15106
ChEBI CHEBI:53452
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 0.1% (w/v)
Chemical Name or Material Naphthylethylenediamine Dihydrochloride
Show More Show Less