CAS RN 14459-95-1
Potassium Ferrocyanide Trihydrate (Crystalline/Certified ACS), Fisher Chemical
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N Synonym: Potassium Prussiate, Yellow,Prussiate of Potash, Yellow IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
ORP Standard, +200 mV, Ricca Chemical
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium hexacyanoferrate(II) trihydrate, ACS, 98.5-102.0%
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium ferrocyanide trihydrate, 99+%, for analysis
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N Synonym: Potassium hexacyanoferrate(II) IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Redox Standard 124 mV, MilliporeSigma™ Supelco™
The value is the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride
Redox Standard 250 mV, MilliporeSigma™ Supelco™
The value is the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride
Potassium Ferrocyanide, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 14459-95-1 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium hexacyanoferrate(II) trihydrate, 98+%
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N Synonym: Potassium ferrocyanide trihydrate IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium Ferrocyanide ACS AR Crystal, Macron Fine Chemicals™
CAS: 14459-95-1 Molecular Formula: K4Fe(CN)6·3H2O
Potassium ferrocyanide trihydrate, 98.5%, ACS reagent
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N Synonym: Potassium hexacyanoferrate(II) IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
ORP Standard, 200mV, Certified, LabChem™
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium Ferrocyanide, Trihydrate, ACS Grade, 98.5 to 102.0%, LabChem™
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium Ferrocyanide Trihydrate ACS MP Biomedicals
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N Synonym: Potassium Hexacyanoferate II IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
Potassium Ferrocyanide, 10% (w/v), Ricca Chemical
CAS: 14459-95-1 Molecular Formula: C6H6FeK4N6O3 Molecular Weight (g/mol): 422.39 MDL Number: MFCD00167023 InChI Key: NLYKFUFIUWPPIZ-UHFFFAOYSA-N IUPAC Name: tetrapotassium hexacyanoironbis(ylium) trihydrate SMILES: O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
- 1
- 2