missing translation for 'onlineSavingsMsg'
Learn More

Carrez I Solution, Fisher Chemical™

Catalog No. SC9101500
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SC9101500 500 mL
SC9101250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SC9101500 Supplier Ricca Chemical Company Supplier No. SC9101500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Carrez I Solution
CAS 14459-95-1, 7732-18-5
CAS Min % 95.0, 3.41
CAS Max % 98.0, 3.55
Grade Laboratory
Packaging Natural Poly Bottle
Methods For AOAC Method 984.15
Quantity 500 mL