missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, Trihydrate, ACS Grade, 98.5 to 102.0%, LabChem™

Catalog No. LC190551
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC190551 500 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC190551 Supplier LabChem Supplier No. LC190551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Yellow
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula K4Fe(CN)6·3H2O
Solubility Information Soluble in water
Formula Weight 422.39
Percent Purity 98.5 to 102.0%
Grade ACS
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.85g/cm3
Decomposition Information Potassium oxide; Carbon monoxide; Carbon dioxide
Density 1.85g/cm3
Chemical Name or Material Potassium Ferrocyanide, Trihydrate
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature