missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide Trihydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. P236500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P236500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P236500 Supplier Thermo Fisher Scientific Supplier No. P236 500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium Prussiate, Yellow, Prussiate of Potash, Yellow
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70°C
Color Yellow
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 98.5 to 102.0%
Linear Formula K4Fe(CN)6·3H2O
Formula Weight 422.39
Percent Purity 98.5 to 102.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 1.8500g/cm³
Chemical Name or Material Potassium Ferrocyanide Trihydrate
Show More Show Less