missing translation for 'onlineSavingsMsg'
Learn More

Redox Standard 250 mV, MilliporeSigma™ Supelco™

Catalog No. 111017268
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017268 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017268 Supplier Merck Emd Millipore Supplier No. 13198500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The value is the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride

Specifications

CAS 14459-95-1
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard