missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, 10% (w/v), Ricca Chemical

Catalog No. 609632
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
609632 1 L Natural Poly Bottle
6096250 250 mL Natural Poly Bottle
609616 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 609632 Supplier Ricca Chemical Company Supplier No. 609632

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Quantity 1 L
Chemical Name or Material Potassium Ferrocyanide, Labeled with manufacturing date