missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK6932212
Change view
Click to view available options
Quantity:
2.5kg
Packaging:
Poly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK6932212 2.5kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK6932212 Supplier Avantor Supplier No. 693205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula K4Fe(CN)6·3H2O

Specifications

Quantity 2.5kg
Formula Weight 422.39
Grade ACS
Packaging Poly
Chemical Name or Material Potassium Ferrocyanide, Trihydrate