CAS RN 127-18-4
Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 IUPAC Name: 1,1,2,2-tetrachloroethene SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, 99%
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, ≥99.5%, Solstice
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 IUPAC Name: tetrachloroethene SMILES: ClC(Cl)=C(Cl)Cl
Density Standard 1623 kg/m3 HD Fitzgerald Ltd. Quality, MilliporeSigma™ Supelco™
Density Standard 1623 kg/m3 also known as tetrachloroethylene is a common industrial chlorinated solvent. It is highly resistant to aerobic biodegradation. It may be a carcinogen and is the most common contaminant in ground water supplies.
Tetrachloroethylene, 99+%, spectrophotometric grade
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, 99%, extra pure
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 IUPAC Name: 1,1,2,2-tetrachloroethene SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, 99+%, for HPLC
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 IUPAC Name: 1,1,2,2-tetrachloroethene SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9%, Solstice
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 SMILES: ClC(Cl)=C(Cl)Cl
Tetrachloroethylene, MilliporeSigma™
CAS: 127-18-4 Molecular Formula: C2Cl4 Molecular Weight (g/mol): 165.82 MDL Number: MFCD00000834 InChI Key: CYTYCFOTNPOANT-UHFFFAOYSA-N Synonym: tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin PubChem CID: 31373 ChEBI: CHEBI:17300 IUPAC Name: tetrachloroethene SMILES: ClC(Cl)=C(Cl)Cl