missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, MilliporeSigma™

Catalog No. M1009652500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1009652500 2.5 L Glass bottle
M1009650500 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1009652500 Supplier Technidata Supplier No. 1.00965.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22°C
Color Colorless
Density 1.62g/cm3
Boiling Point 121°C
Odor Ether-like
Packaging Glass bottle
Quantity 2.5 L
UN Number UN 1897
Solubility Information Water: 0.16g/L at 20°C
Vapor Pressure 19 hPa (20°C)
Formula Weight 165.83
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Hazard Category H351: Suspected of causing cancer.
Hazard Statement H411: Toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 204-825-9
Recommended Storage 15° to 25°C (59° to 77°F); Tightly closed. Protect from heat. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.