missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, 99+%, for HPLC

Catalog No. AC445691000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445691000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445691000 Supplier Thermo Scientific Chemicals Supplier No. 445691000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name 1,1,2,2-tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22.0°C
Density 1.6250g/mL
Boiling Point 120.0°C to 122.0°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.505
Linear Formula Cl2C=CCl2
Quantity 100 mL
Specific Gravity 1.625
Merck Index 15,9335
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene
Viscosity 0.89 mPa.s (20°C)
Formula Weight 165.83
Percent Purity 99+%
Grade HPLC
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Causes serious eye irritati
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 204-825-9

RUO – Research Use Only