missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020681
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020681 1 mL
111019981 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020681 Supplier Merck Emd Millipore Supplier No. 026661ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 127-18-4
Vapor Pressure 19 mmHg (25°C)
Vapor Density 5.83 (vs. air)
Density 1.623 g/mL (at 25°C)
Linear Formula CCl2=CCl2
MDL Number MFCD00000834
Melting Point -22°C
Boiling Point 121°C
Synonym PCE; Perchloroethylene
Type Analytical Standard
Quantity 1 mL
Formula Weight 165.83
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less