missing translation for 'onlineSavingsMsg'
Learn More

Density Standard 1623 kg/m3 HD Fitzgerald Ltd. Quality, MilliporeSigma™ Supelco™

Catalog No. 111018406
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018406 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018406 Supplier Merck Emd Millipore Supplier No. 1588910MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Density Standard 1623 kg/m3 also known as tetrachloroethylene is a common industrial chlorinated solvent. It is highly resistant to aerobic biodegradation. It may be a carcinogen and is the most common contaminant in ground water supplies.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 127-18-4
Vapor Pressure 19 mmHg (25°C)
Vapor Density 5.83 (vs. air)
Density 1.623 g/mL (at 25°C)
Linear Formula CCl2=CCl2
MDL Number MFCD00000834
Melting Point -22°C
Boiling Point 121°C
Synonym Tetrachloroethylene; PCE; Perchloroethylene
Type Organic Standard
Quantity 10 mL
Formula Weight 165.83
Grade H&D; Fitzgerald Ltd. Quality
Show More Show Less