accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 118-56-9

CH3H3CCH3OOOH

CAS RN 118-56-9

IUPAC Name: 3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
Synonyms: coppertone m-homomenthyl salicylate caswell no. 482B homosalato homosalate homosalatum 胡莫柳酯 filtersol ''A'' metahomomenthyl salicylate epa pesticide chemical code 076603
Molecular Weight (g/mol): 262.35
Molecular Formula: C16H22O3
InChi Key: WSSJONWNBBTCMG-UHFFFAOYNA-N
SMILES: CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O
Boiling Point 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.