missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF96 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552315
Description
Immunogen sequence: IIHTGEKPYK CNECGKAFGR WSALNQHQRL HTGEKHYHCN DCGKAFSQKA GLFHHIKIHT RDKPYQCTQC NKSFSRRSIL TQHQGVHTGA KPYECNECGK AFVYNSSLVS HQEIHHK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.
May be involved in transcriptional regulation. Classified as a zinc finger protein expressed in the corpus luteum.Spécifications
| ZNF96 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN12 | |
| 2510038J07Rik; dJ29K1.2; DM9; FPM315; LOW QUALITY PROTEIN: zinc finger and SCAN domain-containing protein 12; mKIAA0426; Zfp96; Zfp-96; zinc finger and SCAN domain containing 12; zinc finger and SCAN domain containing 23; zinc finger and SCAN domain-containing protein 12; zinc finger protein 305; zinc finger protein 323; zinc finger protein 96; ZNF29K1; ZNF305; ZNF323; ZNF96; Zscan12; ZSCAN23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43309 | |
| ZSCAN12 | |
| Recombinant protein corresponding to Human ZNF96. Recombinant protein control fragment (Product #RP-89374). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |