missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Acetate, Crystal, USP, 98-102%, Spectrum™ Chemical
ZI200, 5970-45-6, C4H6O4Zn·2H2O
672,30 $ - 672,30 $
Identifiants chimiques
| CAS | 5970-45-6 |
|---|---|
| Molecular Formula | C4H10O6Zn |
| Molecular Weight (g/mol) | 219.50 |
| InChI Key | BEAZKUGSCHFXIQ-UHFFFAOYSA-L |
| IUPAC Name | zinc(2+) diacetate dihydrate |
| SMILES | O.O.[Zn++].CC([O-])=O.CC([O-])=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
18607813
|
Spectrum Chemical Mfg Cor
ZI200500GM |
500 g | Poly Bottle |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Spectrum™ Chemical Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Identifiants chimiques
| 5970-45-6 | |
| 219.50 | |
| zinc(2+) diacetate dihydrate |
| C4H10O6Zn | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| O.O.[Zn++].CC([O-])=O.CC([O-])=O |
Spécifications
| 5970-45-6 | |
| 6.0 to 8.0 | |
| C4H10O6Zn | |
| O.O.[Zn++].CC([O-])=O.CC([O-])=O | |
| 219.50 | |
| USP |
| 1 | |
| 500 g | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| zinc(2+) diacetate dihydrate | |
| 98 to 102% | |
| Poly Bottle |