missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, Crystal, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18607813
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607813 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607813 Supplier Spectrum Chemical Mfg Cor Supplier No. ZI200500GM
Only null left

ZI200, 5970-45-6, C4H6O4Zn·2H2O

Spectrum™ Chemical Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.50
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
IUPAC Name zinc(2+) diacetate dihydrate
SMILES O.O.[Zn++].CC([O-])=O.CC([O-])=O

Specifications

pH 6.0 to 8.0
CAS Max % 1
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Packaging Poly Bottle