missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFHX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149963
Description
ZFHX4 Polyclonal Antibody for Western Blot, ICC/IF, IHC
May play a role in neural and muscle differentiation. May be involved in transcriptional regulation.Spécifications
| ZFHX4 | |
| Polyclonal | |
| Unconjugated | |
| ZFHX4 | |
| A930021B15; C130041O22Rik; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; RGD1563022; ZFH4; Zfh-4; Zfhx4; ZHF4; zinc finger homeobox 4; zinc finger homeobox protein 4; zinc finger homeobox protein 4; LOW QUALITY PROTEIN: zinc finger homeobox protein 4; zinc finger homeodomain 4; zinc finger homeodomain protein 4; zinc-finger homeodomain protein 4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 310250, 79776, 80892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q86UP3, Q9JJN2 | |
| ZFHX4 | |
| Recombinant Zinc Finger Homeobox Protein 4 (ZFHX4) (Met1-Pro189). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |