missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ WNT3 (Human) Recombinant Protein
Human WNT3 full-length ORF ( NP_110380.1, 1 a.a. - 355 a.a.) recombinant protein with GST-tag at N-terminal.
Supplier: Abnova™ H00007473P012
Description
- Sequence: MEPHLLGLLLGLLLGGTRVLAGYPIWWSLALGQQYTSLGSQPLLCGSIPGLVPKQLRFCRNYIEIMPSVAEGVKLGIQECQHQFRGRRWNCTTIDDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTSTICGCDSHHKGPPGEGWKWGGCSEDADFGVLVSREFADARENRPDARSAMNKHNNEAGRTTILDHMHLKCKCHGLSGSCEVKTCWWAQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRAKYSLFKPPTERDLVYYENSPNFCEPNPETGSFGTRDRTCNVTSHGIDGCDLLCCGRGHNTRTEKRKEKCHCIFHWCCYVSCQECIRIYDVHTCK
Spécifications
| NP_110380.1 | |
| wingless-type MMTV integration site family, member 3 | |
| 125% SDS-PAGE Stained with Coomassie Blue | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| WNT3 | |
| GST |
| 7473 | |
| Wheat germ expression system | |
| 2 μg | |
| INT4, MGC131950, MGC138321, MGC138323 | |
| Wheat Germ (in vitro) | |
| 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |