Learn More
DWK Life Sciences Wheaton™ Trypsinizing Flasks with Side Arm
Flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents.
$246.60 - $355.50
Spécifications
Description Style: Erlenmeyer Trypsinizing
Material Borosilicate Glass
For Use With (Application) Used for forming cell suspensions through trypsin digestion of connective proteins
Includes 20-415 Sidearm cap and top cap have white Styrene Butadiene Rubber liner
Autoclavable Autoclavable
voir les spécifications
Produits 5
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Graduated Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Graduated Prix Quantité  
10-042-11A
Afficher les documents
DWK Life Sciences
355752
35 mL No
caisse for $286.07
Il en reste null
10-042-11D
Afficher les documents
DWK Life Sciences
355755
300 mL No
chaque for $325.33
Il en reste null
10-042-12B
Afficher les documents
DWK Life Sciences
355803
75 mL Yes
caisse for $355.50
Il en reste null
10-042-11C
Afficher les documents
DWK Life Sciences
355754
150 mL No
caisse for $268.83
Il en reste null
10-042-11B
Afficher les documents
DWK Life Sciences
355753
75 mL No
caisse for $246.60
Il en reste null
Description
Description
  • Sidearm allows easier decanting of contents while retaining large particles behind the dam
  • Sidearm caps are 20-415 size
  • Caps have white styrene-butadiene rubber liners
  • Flasks made from Wheaton 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E- 438 Type I, Class A requirements
Spécifications
Spécifications
Style: Erlenmeyer Trypsinizing
Used for forming cell suspensions through trypsin digestion of connective proteins
Autoclavable
Borosilicate Glass
20-415 Sidearm cap and top cap have white Styrene Butadiene Rubber liner
Screw
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'