Learn More
DWK Life Sciences Wheaton™ Polycarbonate Erlenmeyer Shake Flasks
Ideal replacement for glass Erlenmeyer/Shake Flasks
Supplier: DWK Life Sciences WPFPC0500S
Description
- Made of Lexan Polycarbonate Resin that has mechanical, optical, electrical and thermal properties
- Supplied standard with vented, autoclavable proprietary DuoCap™ that allows gas exchange
- Molded in graduation markings
- Certified pyrogen, DNase and RNase free
- Gamma irradiated to sterility assurance level (SAL) of 10-6
- Sterile flat base flasks individually wrapped in easy to tear sterile bag
- Flame retardant and scratch resistant resin meets USP requirements
Spécifications
| Blue | |
| Clear | |
| Polycarbonate | |
| Vented DuoCap |
| Polypropylene | |
| 500 mL | |
| 12/Pk. | |
| 38-430 |