Learn More
DWK Life Sciences™ Wheaton™ KeepIT™ Freezer Boxes
Provides an ideal method for batching and storing samples.
Supplier:  DWK Life Sciences™ W651704W
 Afficher plus de versions de ce produit
Numéro de catalogue. 13-606-108
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Six different colors match the colors of CryoELITE Cryogenic Vials, creating alternatives for batching and identifying groups of samples.
  • KeepIT-100 accommodates up to 100 internal threaded vials, KeepIT-81 accommodates 81 external threaded vials, and KeepIT-25 accommodates up to 25 internal or external threaded cryogenic vials; Vial sizes: 1.2 – 2mL
  • Openings in bottom facilitate scanning CryoELITE 2D Data Matrix Bar Code Inserts and allow for drainage of liquid nitrogen
  • Made from Eastman Tritan™ BPA-free, shatter-resistant resin
  • Standard footprint is compatible with liquid nitrogen storage shelves and freezer drawers
Spécifications
Spécifications
White
Square
Batching and Storing Samples
5.1 x 5.1 x 2 in. (13 x 13 x 5.2 cm)
13 cm
13 cm
5.2 cm
Copolyester Resin
Freezer Box
100 (1.2 to 2 mL Internal Threaded Cryogenic Vial)
5.1 in.
5.1 in.
2 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS