Learn More
DWK Life Sciences™ Wheaton™ KeepIT™ Freezer Boxes
Provides an ideal method for batching and storing samples.
$302.67 - $383.70
Specifications
| Dimensions (L x W x H) | 5.1 x 5.1 x 2 in. (13 x 13 x 5.2 cm) |
|---|---|
| Length (English) | 5.1 in. |
| Length (Metric) | 13 cm |
| Width (English) | 5.1 in. |
| Width (Metric) | 13 cm |
| Catalog Number | Mfr. No. | Color | Holds | Type | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Holds | Type | Price | Quantity | |||
|
13-606-105
|
DWK Life Sciences™
W651703B |
Blue | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
03-084-102
|
DWK Life Sciences™
W651702R |
Red | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-102
|
DWK Life Sciences™
W651703W |
White | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
03-084-106
|
DWK Life Sciences™
W651702B |
Blue | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-101
|
DWK Life Sciences™
W651703P |
Pink | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $323.25
|
|
|||
|
13-606-104
|
DWK Life Sciences™
W651703G |
Green | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
13-606-106
|
DWK Life Sciences™
W651704R |
Red | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $323.25
|
|
|||
|
13-606-103
|
DWK Life Sciences™
W651703Y |
Yellow | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $323.25
|
|
|||
|
03-084-105
|
DWK Life Sciences™
W651702G |
Green | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-100
|
DWK Life Sciences™
W651703R |
Red | 81 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
03-084-104
|
DWK Life Sciences™
W651702Y |
Yellow | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-107
|
DWK Life Sciences™
W651704P |
Pink | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
03-084-101
|
DWK Life Sciences™
W651702W |
White | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-109
|
DWK Life Sciences™
W651704Y |
Yellow | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
13-606-108
|
DWK Life Sciences™
W651704W |
White | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
13-606-111
|
DWK Life Sciences™
W651704B |
Blue | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $302.67
|
|
|||
|
03-084-103
|
DWK Life Sciences™
W651702P |
Pink | 25 (1.2 to 2 mL External Threaded Cryogenic Vial) | Freezer Box |
Case of 10 Each for $383.70
|
|
|||
|
13-606-110
|
DWK Life Sciences™
W651704G |
Green | 100 (1.2 to 2 mL Internal Threaded Cryogenic Vial) | KeepIT -100 Freezer Box |
Case of 10 Each for $302.67
|
|
|||
Description
Six different colors match the colors of CryoELITE Cryogenic Vials, creating alternatives for batching and identifying groups of samples.
- KeepIT-100 accommodates up to 100 internal threaded vials, KeepIT-81 accommodates 81 external threaded vials, and KeepIT-25 accommodates up to 25 internal or external threaded cryogenic vials; Vial sizes: 1.2 – 2mL
- Openings in bottom facilitate scanning CryoELITE 2D Data Matrix Bar Code Inserts and allow for drainage of liquid nitrogen
- Made from Eastman Tritan™ BPA-free, shatter-resistant resin
- Standard footprint is compatible with liquid nitrogen storage shelves and freezer drawers
Specifications
| 5.1 x 5.1 x 2 in. (13 x 13 x 5.2 cm) | |
| 13 cm | |
| 13 cm | |
| 5.2 cm |
| 5.1 in. | |
| 5.1 in. | |
| 2 in. |