missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149836
Description
VWA2 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the von Willebrand factor A-like domain protein superfamily. The encoded protein is localized to the extracellular matrix and may serve as a structural component in basement membranes or in anchoring structures on scaffolds of collagen VII or fibrillin. This gene has been linked to type 1A diabetes and is a candidate serological marker for colon cancer.Spécifications
| VWA2 | |
| Polyclonal | |
| Unconjugated | |
| VWA2 | |
| a domain-containing protein similar to matrilin and collagen; AMACO; CCSP-2; colon cancer diagnostic marker; colon cancer secreted protein 2; NET42; von Willebrand factor A domain containing 2; von Willebrand factor A domain-containing protein 2; VWA2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100152718, 340706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5GFL6 | |
| VWA2 | |
| Recombinant Von Willebrand Factor A Domain Containing Protein 2 (vWA2) (Ser457-Gly708). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody |