missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoir Cooler
Custom-fit companion for VistaLab 100mL reagent reservoirs
Supplier: VistaLab™ Technologies 30545000
Comprend: Three 100mL reservoirs and lids.
Description
- Reusable cooler maintains fluid temperatures at less than 5°C for up to two hours
- Constructed of heavy-duty PVC that encases a quick-freezing, nontoxic bright blue gel
- Provides consistent cooling without contamination risk or inconvenience of ice baths
Spécifications
| Blue | |
| PVC | |
| Reservoir Cooler | |
| 6 in. | |
| 3/Pk. | |
| 8 in. | |
| 1 | |
| V-bottom |
| 100 mL | |
| Non-sterile | |
| General Liquid Handling | |
| 10 in. | |
| 100 mL | |
| Not Autoclavable | |
| Reusable |