Learn More
Wells Lamont™ Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves
Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves are constructed with a unique blend of high performance fibers and stainless steel to provide comfort and cut resistance. Polyurethane coating on the palm enhances grip capabilities when handling small parts.
Supplier:  Wells Lamont™ Y9294XS
 Afficher plus de versions de ce produit
Numéro de catalogue. 19048784
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Constructed from a blend of high performance fibers and stainless steel
  • 13-gauge shell provides ANSI Level A4 cut protection
  • Polyurethane coating on the palm enhances grip and tactility
  • Hi-vis coloring increases visibility during low light applications
  • Open, uncoated back circulates air to keep hands cool and comfortable
Spécifications
Spécifications
ANSI 5
Polyurethane
ANSI A4
HPPE, Stainless Steel Fiber
X-Small
Assembly, construction, metal fabrication
Y9294XS
13
ASTM F1790-97, ASTM F2992-15, ASTM D3889-10
Hi-vis Orange, Black
Specific
Cut-resistant Gloves
EN 388:4543
Knit
FlexTech™ Vis-Tech Y9294
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS