Learn More
Wells Lamont™ Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves
Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves are constructed with a unique blend of high performance fibers and stainless steel to provide comfort and cut resistance. Polyurethane coating on the palm enhances grip capabilities when handling small parts.
$205.79 - $205.79
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance ASTM F1790-97, ASTM F2992-15, ASTM D3889-10
Coating Material Polyurethane
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A4
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19048787
View Documents
Wells Lamont™
Y9294L
Large
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
19048786
View Documents
Wells Lamont™
Y9294M
Medium
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
19048785
View Documents
Wells Lamont™
Y9294S
Small
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
19048784
View Documents
Wells Lamont™
Y9294XS
X-Small
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
19048788
View Documents
Wells Lamont™
Y9294XL
X-Large
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
19048789
View Documents
Wells Lamont™
Y9294XXL
2X-Large
Dozen of 12 for $205.79
Case of 12 Dozen for $2,116.80
Only null left
Description
Description
  • Constructed from a blend of high performance fibers and stainless steel
  • 13-gauge shell provides ANSI Level A4 cut protection
  • Polyurethane coating on the palm enhances grip and tactility
  • Hi-vis coloring increases visibility during low light applications
  • Open, uncoated back circulates air to keep hands cool and comfortable
Specifications
Specifications
ANSI 5
Polyurethane
ANSI A4
HPPE, Stainless Steel Fiber
EN 388:4543
Knit
13
ASTM F1790-97, ASTM F2992-15, ASTM D3889-10
Hi-vis Orange, Black
Specific
Cut-resistant Gloves
Assembly, construction, metal fabrication
FlexTech™ Vis-Tech Y9294
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'