missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ 3.5 in. Pipe Brush for Flex Rod

Vikan™ 3.5 in. Pipe Brush for Flex Rod is designed to handle scrubbing while still pick up fine particles.

Supplier:  Vikan™ 53914

 Afficher plus de versions de ce produit

Numéro de catalogue. 17-227-464


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Ideal for cleaning machinery and equipment with narrow valves, pipes, and other parts
  • This full-bristle tube brush cleans many commonly sized tubes and valves
  • The specialized thread on this brush fits the Vikan™ long flexible handles 5350 and 53525
  • The bristles are dense and firm enough to clean thoroughly, but flexible enough to move with the shape of the equipment being cleaned
  • Color-coded
Spécifications

Spécifications

Pipe Brush
Medium Stiffness
Stainless Steel
FDA Compliant
3.5 in.
1 Ea.
Polyester
Yes
Non-sterile
Red
7.7 in.
Medium
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions