missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ViiA™ 7 Dx Instrument, Array Card
With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, available for in-vitro diagnostics use.
$99,300.00 - $118,800.00
Spécifications
| Block Format | Interchangeable |
|---|---|
| Dynamic Range | Up to 9 Logs of Linear Dynamic Range |
| Thermal Uniformity | ±0.5°C |
| Voltage | 100 to 240 V |
| Height (Metric) | 64.5 cm |
| Numéro de catalogue | Numéro du manufacturier. | Content And Storage | Description | Volume (Metric) Thermal Block Sample | Prix | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Content And Storage | Description | Volume (Metric) Thermal Block Sample | Prix | Quantity & Availability | |||
4453534
|
Applied Biosystems™
4453534 |
96-well Block | Scan Time: About 35 min. (varies depending on reagents and other factors); Power limit: 60Hz | 10 to 100μL |
N/A
|
N/A | |||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
4453535
|
Applied Biosystems™
4453535 |
Fast 96-well Block | Scan Time: About 35 min. (varies depending on reagents and other factors); Power limit: 60Hz | 10 to 30μL |
N/A
|
N/A | |||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
4453537
|
Applied Biosystems™
4453537 |
TaqMan 384-well Array Block | Scan Time: About 35 min. (varies depending on reagents and other factors); Power limit: 60Hz | 1μL |
N/A
|
N/A | |||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
4453536
|
Applied Biosystems™
4453536 |
384-well Block | Scan Time: About 35 min. (varies depending on reagents and other factors); Power limit: 60Hz | 5 to 20μL |
N/A
|
N/A | |||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
With a simplified workflow, intuitive software, touch-screen interface, and one-button protocols for error minimization, the ViiA™ 7 System offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Full com
Key product features:- Proven Applied Biosystems reliability and accuracy
- Using with Micro Fluidic Array Cards provides an integrated workflow and faster results
- Easy to use - intuitive software, responsive touch-scre
Spécifications
| Interchangeable | |
| ±0.5°C | |
| 64.5 cm | |
| 4°C to 100°C | |
| Scan Time: About 35 min. (varies depending on reagents and other factors); Power limit: 60Hz | |
| 6 Emission Filters (500 to 720 nm), 6 Excitation Filters (450 to 670 nm), OptiFlex™ system (Enhanced Fluorescence Detection) | |
| 60.7 kg (134 lb.) |
| Up to 9 Logs of Linear Dynamic Range | |
| 100 to 240 V | |
| 63.5 cm | |
| Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling | |
| High-throughput Compatible, Multiplexing | |
| 1 System |