missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Thallium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010630100N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Spécifications
| Thallium ICP-MS Standard | |
| 10102-45-1,7697-37-2,7732-18-5 | |
| 99.96%,2.04%,0.00% | |
| Liquid | |
| MFCD00011280 | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| [Tl+].[O-][N+]([O-])=O | |
| 266.38 | |
| 24937 |
| Natural LDPE Bottle | |
| 96.04%,1.96%,0.00% | |
| 1 | |
| NO3Tl | |
| Colorless | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| FYWSTUCDSVYLPV-UHFFFAOYSA-N | |
| λ1-thallanylium nitrate | |
| 100 mL | |
| High Purity |
Identifiants chimiques
| 10102-45-1 | |
| 266.38 | |
| FYWSTUCDSVYLPV-UHFFFAOYSA-N | |
| λ1-thallanylium nitrate |
| NO3Tl | |
| MFCD00011280 | |
| 24937 | |
| [Tl+].[O-][N+]([O-])=O |