Learn More
Bolle Safety™ Vented Universal Safety Goggles
Numéro de catalogue. 17467448
Change view
missing translation for 'orderingAttributeHoverText'
Style:
Clear
Clear and Foam
Clear, Foam, and Mouthguard
Smoke and Foam
Numéro de catalogue. Style
17467448 Clear, Foam, and Mouthguard
17467444 Smoke and Foam
17467446 Clear and Foam
17467451 Clear
4 options
Numéro de catalogue. 17467448
Supplier: Bolle Safety™ UNIVGN14W
Il en reste null
This goggle provides optimum protection and comfort in all circumstances thanks to its ergonomic design. This innovative goggle with its exclusive platinum coating is available in several versions, each adapted to a specific work environment to offer more peace of mind and efficiency for the worker.
  • Optimal fit thanks to its Wave technology
  • PC/TPR co-injected frame with overflow chute
  • Effortless pivoting faceguard
  • Improved recyclability
  • UV protection
  • Scratch-resistant
  • Anti-fog
Spécifications
Lens Coating Scratch-resistant, Anti-fog
Description Vented
Color Black and Yellow
Lens Tint Clear
Product Type Safety Goggles
Style Clear, Foam, and Mouthguard
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Standard
Afficher plus de résultats Afficher moins de résultats