Learn More
Thermo Scientific Chemicals USA Standard Sieve, 100 Mesh
Brass frame and Stainless-steel mesh sieves
$206.81 - $287.64
Spécifications
Description 100 Mesh
Type Sieve
Certifications/Compliance ASTM E11
Frame Material Brass
Mesh Material Stainless Steel
voir les spécifications
Produits 2
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Outer Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Outer Prix Quantité  
AA39987ON
Afficher les documents
Thermo Scientific Chemicals
039987ON
7.62 cm
chaque for $206.81
Il en reste null
AA39987NR
Afficher les documents
Thermo Scientific Chemicals
039987NR
20.3 cm
chaque for $287.64
Il en reste null
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep
Spécifications
Spécifications
100 Mesh
ASTM E11
Stainless Steel
Sieve
Brass
100
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'
RUO – Research Use Only