Learn More
DuPont™ Tyvek™ IsoClean™ 265 Series Lab Coats
Tyvek™ IsoClean™ lab coats provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in controlled environments.
Supplier: DuPont™ IC265SWHXL00300B
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Knit cuff for tighter fit at wrist.
- Front zipper closure for easy donning and doffing.
- Raglan sleeve design for greater range of motion and comfort.
- Three front pockets.
- A-line silhouette.
- Bulk packaged in double transparent poly liners.
- Manufactured under specifications that do not contain natural rubber latex
- D15541914,D15541920,D15541915,D15541916,D15541917,D15541918,D15541919
Spécifications
| Controlled Environments Lab Coat | |
| Unisex | |
| Serged | |
| Bulk Packaged, Double Transparent Poly Liners |
| X-Large | |
| Tyvek | |
| Raglan |