Learn More
TrippNT Extra Large Draft Shield
TrippNT 50240 Extra Large Acrylic Draft Shield. Protects balances and delicate instruments from drafts. See through with cut out for threading cords or wires and one side open for access. 22x22x22 inches (width height depth) 17 pounds
Supplier: TrippNT TRP50240
Description
TrippNT 50240 Extra Large Acrylic Draft Shield. Protects balances and delicate instruments from drafts. See through with cut out for threading cords or wires and one side open for access. 22x22x22 inches (width height depth) 17 pounds
Spécifications
| Clear | |
| 22 x 22 x 22 in. | |
| 22 in. | |
| Overhanging panel for protection from downward drafts; Half circle cut out at bottom fits cords or wires |
| Acrylic | |
| 22 in. | |
| 22 in. | |
| Draft Shield |