missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-10-Hydroxy-2-decenoic Acid 97.0+%, TCI America™
Supplier: TCI America H1337200MG
Spécifications
| trans-10-Hydroxy-2-decenoic Acid | |
| 66°C | |
| C10H18O3 | |
| 200 mg | |
| OCCCCCCC\C=C\C(O)=O | |
| 186.25 | |
| CHEBI:78668 | |
| ≥97.0% (GC,T) |
| 14113-05-4 | |
| White | |
| MFCD00204506 | |
| QHBZHVUGQROELI-SOFGYWHQSA-N | |
| (2E)-10-hydroxydec-2-enoic acid | |
| 5312738 | |
| 186.25 | |
| Crystalline Powder |
Identifiants chimiques
| 14113-05-4 | |
| 186.25 | |
| QHBZHVUGQROELI-SOFGYWHQSA-N | |
| CHEBI:78668 | |
| OCCCCCCC\C=C\C(O)=O |
| C10H18O3 | |
| MFCD00204506 | |
| 5312738 | |
| (2E)-10-hydroxydec-2-enoic acid |
Sécurité et manipulation
missing translation for 'einecsNumber' : (2)-1340
missing translation for 'tsca' : No
Recommended Storage : Refrigerator