missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-10-Hydroxy-2-decenoic Acid 97.0+%, TCI America™
$126.76 - $445.51
Chemical Identifiers
| CAS | 14113-05-4 |
|---|---|
| Molecular Formula | C10H18O3 |
| Molecular Weight (g/mol) | 186.25 |
| MDL Number | MFCD00204506 |
| InChI Key | QHBZHVUGQROELI-SOFGYWHQSA-N |
| PubChem CID | 5312738 |
| ChEBI | CHEBI:78668 |
| IUPAC Name | (2E)-10-hydroxydec-2-enoic acid |
| SMILES | OCCCCCCC\C=C\C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
H1337200MG
|
TCI America
H1337200MG |
200 mg |
Each for $126.76
|
|
|||||
|
H13371G
|
TCI America
H13371G |
1 g |
Each for $445.51
|
|
|||||
Chemical Identifiers
| 14113-05-4 | |
| 186.25 | |
| QHBZHVUGQROELI-SOFGYWHQSA-N | |
| CHEBI:78668 | |
| OCCCCCCC\C=C\C(O)=O |
| C10H18O3 | |
| MFCD00204506 | |
| 5312738 | |
| (2E)-10-hydroxydec-2-enoic acid |
Specifications
| 14113-05-4 | |
| White | |
| MFCD00204506 | |
| QHBZHVUGQROELI-SOFGYWHQSA-N | |
| (2E)-10-hydroxydec-2-enoic acid | |
| 5312738 | |
| 186.25 | |
| Crystalline Powder |
| 66°C | |
| C10H18O3 | |
| 200 mg | |
| OCCCCCCC\C=C\C(O)=O | |
| 186.25 | |
| CHEBI:78668 | |
| ≥97.0% (GC,T) | |
| trans-10-Hydroxy-2-decenoic Acid |
Safety and Handling
EINECSNumber : (2)-1340
TSCA : No
Recommended Storage : Refrigerator