missing translation for 'onlineSavingsMsg'
Learn More

Thorium Standard For ICP/MS, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLTH21BY

Numéro de catalogue. CLTH21BY


Il en reste null
Découvrez plus d'offres spéciales

Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Th1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Spécifications

Thorium Standard
7440-29-1
Th
ZSLUVFAKFWKJRC-UHFFFAOYSA-N
2% HNO3
232.038
23960
1mg/L
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
[Th]
thorium
125 mL
CHEBI:33385

Identifiants chimiques

7440-29-1
232.038
23960
thorium
Th
ZSLUVFAKFWKJRC-UHFFFAOYSA-N
CHEBI:33385
[Th]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

FDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only