Learn More
Thiocarbohydrazide, 97%
CAS: 2231-57-4 | CH6N4S | 106.15 g/mol
87,38 $ - 889,71 $
Identifiants chimiques
| CAS | 2231-57-4 |
|---|---|
| Molecular Formula | CH6N4S |
| Molecular Weight (g/mol) | 106.15 |
| MDL Number | MFCD00007616 |
| InChI Key | LJTFFORYSFGNCT-UHFFFAOYSA-N |
| Synonym | thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu |
| PubChem CID | 2724189 |
| ChEBI | CHEBI:9549 |
| IUPAC Name | 1,3-diaminothiourea |
| SMILES | NNC(=S)NN |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAL0120506
|
Thermo Scientific Chemicals
L0120506 |
5 g |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAL0120514
|
Thermo Scientific Chemicals
L0120514 |
25 g |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAL0120522
|
Thermo Scientific Chemicals
L0120522 |
100 g |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.
Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.
Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
Identifiants chimiques
| 2231-57-4 | |
| 106.15 | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 2724189 | |
| 1,3-diaminothiourea |
| CH6N4S | |
| MFCD00007616 | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| CHEBI:9549 | |
| NNC(=S)NN |
Spécifications
| 2231-57-4 | |
| CH6N4S | |
| 5 g | |
| 506657 | |
| thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu | |
| LJTFFORYSFGNCT-UHFFFAOYSA-N | |
| 1,3-diaminothiourea | |
| 2724189 | |
| 106.15 | |
| Thiocarbohydrazide |
| ∽175°C (decomposition) | |
| MFCD00007616 | |
| UN2811 | |
| Light sensitive | |
| Very slightly soluble (0.5g/100g,25°C). Insoluble in ether. | |
| NNC(=S)NN | |
| 106.15 | |
| CHEBI:9549 | |
| 97% |
Sécurité et manipulation
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H300+H330-H311
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 218-769-8
missing translation for 'rtecsNumber' : FF2975000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only