missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbohydrazide, 97%

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol

Supplier:  Thermo Scientific Chemicals L0120514

 View more versions of this product

Catalog No. AAL0120514


Only null left
Explore available promotions

Description

Description

Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.

Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
Specifications

Specifications

Thiocarbohydrazide
∽175°C (decomposition)
MFCD00007616
UN2811
Light sensitive
Very slightly soluble (0.5g/100g,25°C). Insoluble in ether.
NNC(=S)NN
106.15
CHEBI:9549
97%
2231-57-4
CH6N4S
25 g
506657
thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
LJTFFORYSFGNCT-UHFFFAOYSA-N
1,3-diaminothiourea
2724189
106.15

Chemical Identifiers

2231-57-4
106.15
LJTFFORYSFGNCT-UHFFFAOYSA-N
2724189
1,3-diaminothiourea
CH6N4S
MFCD00007616
thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
CHEBI:9549
NNC(=S)NN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c

H300+H330-H311

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 218-769-8

RTECSNumber : FF2975000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only