missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, 10% in Water

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

$125.84 - $492.87

Spécifications

Chemical Name or Material Tetraethylammonium hydroxide
Color Colorless
Name Note 10% in Water
CAS 77-98-5 , 7732-18-5
Boiling Point 102.0°C
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AC420281000
Afficher les documents
Thermo Scientific Chemicals
420281000
100 g
chaque for $125.84
Only null left
 
AC420285000
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
420285000
500 g
chaque for $492.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetraethylammonium hydroxide
10% in Water
102.0°C
(C2H5)4NOH
1.0200g/mL
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
9.5 to 10.5%
1.402
Liquid
Colorless
77-98-5 , 7732-18-5
C8H21NO
MFCD00009024
15, 9346
Solubility in water: soluble.
[OH-].CC[N+](CC)(CC)CC
147.26
147.26
Plastic bottle
1.02
Videos
SDS
Documents

Documents

RUO – Research Use Only