missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Dibromoiodide 97.0+%, TCI America™
Supplier: TCI America T126910G
Spécifications
| Tetrabutylammonium Dibromoiodide | |
| 81°C | |
| MFCD00059121 | |
| LFZGDXLUTJIZAG-UHFFFAOYSA-N | |
| 529.183 | |
| 529.18 | |
| Crystalline Powder |
| 15802-00-3 | |
| C16H36Br2IN | |
| 10 g | |
| CCCC[N+](CCCC)(CCCC)CCCC.Br[I-]Br | |
| 13867581 | |
| ≥97.0% (T) |
Identifiants chimiques
| 15802-00-3 | |
| 529.183 | |
| LFZGDXLUTJIZAG-UHFFFAOYSA-N | |
| CCCC[N+](CCCC)(CCCC)CCCC.Br[I-]Br |
| C16H36Br2IN | |
| MFCD00059121 | |
| 13867581 |
Sécurité et manipulation
missing translation for 'einecsNumber' : (2)-0186
missing translation for 'tsca' : No