missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Dibromoiodide 97.0+%, TCI America™

Catalog No. p-7136689
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12691G 1 g
T126910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12691G Supplier TCI America Supplier No. T12691G
Only null left

Chemical Identifiers

CAS 15802-00-3
Molecular Formula C16H36Br2IN
Molecular Weight (g/mol) 529.183
MDL Number MFCD00059121
InChI Key LFZGDXLUTJIZAG-UHFFFAOYSA-N
PubChem CID 13867581
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Br[I-]Br

Specifications

Melting Point 81°C
Quantity 1 g
Formula Weight 529.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Dibromoiodide
EINECSNumber (2)-0186
TSCA No