Learn More
Synthware™ Vacuum or Argon Adapters
With hose connection on the side and male joint
Supplier: Synthware™ A292942
Description
Synthware vacuum or inert gas adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection is at a 90 degree angle to the standard taper inner joint.Spécifications
| Organic Synthesis | |
| Vacuum or Argon Adapter | |
| 29/42 |
| Borosilicate Glass (3.3) | |
| 1 |