Learn More
Synthware™ Vacuum or Argon Adapters
With hose connection on the side and male joint
$45.77 - $50.67
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass (3.3) |
| Product Type | Vacuum or Argon Adapter |
| Quantity | 1 |
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
|
31500531
|
Synthware™
A291420 |
14/20 |
Each for $45.77
|
|
|||||
|
31500532
|
Synthware™
A291922 |
19/22 |
Each for $47.53
|
|
|||||
|
31500533
|
Synthware™
A292440 |
24/40 |
Each for $49.33
|
|
|||||
|
31500534
|
Synthware™
A292942 |
29/42 |
Each for $50.67
|
|
|||||
Description
Synthware vacuum or inert gas adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection is at a 90 degree angle to the standard taper inner joint.Specifications
| Organic Synthesis | |
| Borosilicate Glass (3.3) | |
| Vacuum or Argon Adapter | |
| 1 |