Learn More
Synthware™ Chromatography Column with Spherical Joint, PTFE stopcock and Fritted Disc
General purpose chromatography column for flash chromatography
Supplier:  Synthware™ C195264C
 Afficher plus de versions de ce produit
Numéro de catalogue. 31500933
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Equipped with PTFE stopcock and top 35/20 socket joint
  • With coarse porosity fritted disc to support packing material
  • Synthware columns designed with heavy-walls for increased safety
Spécifications
Spécifications
20 mm
35/20
457 mm
Spherical joint
2 mm bore
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS