Learn More
(-)-Strychnine, 98%
CAS: 57-24-9 | C21H22N2O2 | 334.42 g/mol
Supplier: Thermo Scientific Chemicals 158950250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| (-)-Strychnine | |
| 57-24-9 | |
| 100.0 | |
| White | |
| 98% | |
| C21H22N2O2 | |
| 25 g | |
| strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos | |
| QMGVPVSNSZLJIA-FVWCLLPLSA-N | |
| (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one | |
| 441071 | |
| 334.41 | |
| Fine Crystalline Powder |
| 98% | |
| 97.5 | |
| 284.0°C to 286.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005941 | |
| 15, 8985 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in chlororform and hot ethanol,insoluble in diethyl ether | |
| O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61 | |
| 334.42 | |
| CHEBI:28973 | |
| 98% |
Identifiants chimiques
| 57-24-9 | |
| 334.42 | |
| QMGVPVSNSZLJIA-FVWCLLPLSA-N | |
| 441071 | |
| (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one |
| C21H22N2O2 | |
| MFCD00005941 | |
| strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos | |
| CHEBI:28973 | |
| O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61 |
Sécurité et manipulation
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-319-7
missing translation for 'rtecsNumber' : WL2275000
missing translation for 'tsca' : TSCA
RUO – Research Use Only